Order Entry
Belgium
Orders LinkContactUsLinkComponent
RIPA cell lysis buffer 2

RIPA cell lysis buffer 2

RIPA cell lysis buffer 2, 100 mL
Catalog # ENZOADI801284
Supplier:   Enzo Life Sciences
RIPA cell lysis buffer 2
Catalog # ENZOADI801284
Supplier:  Enzo Life Sciences
CAS Number:  
RIPA cell lysis buffer 2

RIPA cell lysis buffer 2, 100 mL

CAS Number:  
Price
322,00 €
Log in for availability
1 * 100 mL
Product Details & Documents
Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    RIPA cell lysis buffer 2
  • Cat. No.:
    ENZOADI801284
  • Formulation:
    Liquid, 50 mm TRIS-hydrogen chloride, pH 7,4, containing 150 mm sodium chloride, 1 mm EDTA, 1 mm EGTA, 1% Triton X-100, 1% sodium deoxycholate and 0,1% SDS

Specifications