Order Entry
Belgium
Orders LinkContactUsLinkComponent
IHC Wash Buffer, Enzo Life Sciences

IHC Wash Buffer, Enzo Life Sciences

IHC wash buffer, Enzo life sciences, 1 × 1 L, 1.000 mL
Catalog # ENZOADI9502351000
Supplier:   Enzo Life Sciences
IHC Wash Buffer, Enzo Life Sciences
Catalog # ENZOADI9502351000
Supplier:  Enzo Life Sciences
CAS Number:  
IHC Wash Buffer, Enzo Life Sciences

IHC wash buffer, Enzo life sciences, 1 × 1 L, 1.000 mL

CAS Number:  
Price
170,00 €
Log in for availability
1 * 1.000 mL
Product Details & Documents

The IHC wash buffer is a for washing and removing excess reagents during immunostaining.

Liquid. 10X wash buffer.

Specifications

Specifications

  • Pk:
    1.000 mL
  • Description:
    IHC wash buffer, Enzo life sciences
  • Pk Info:
    1 × 1 L

Specifications