Order Entry
Belgium
ContactUsLinkComponent
 
In-situ hybridisation buffer (1.25X concentrate), Enzo Life Sciences
Catalog # ENZOENZ33808
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
In-situ hybridisation buffer (1.25X concentrate), Enzo Life Sciences
Catalog # ENZOENZ33808
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Description:
    In situ 1,25X
  • Seq:
    02
  • Formulation:
    Liquid. Sodium phosphate, NaCl and EDTA (SSPE) buffered formamide, containing hybridisation enhancers.

Specifications

About this item

In Situ Hybridization Buffer provides a high quality hybridization medium for in situ hybridization analyses using either DNA or oligonucleotide probes labeled with biotin-, digoxigenin- or fluorescein-modified nucleotides.

Ordering information: Quantity: Provides a sufficient amount of hybridization buffer concentrate to prepare 12.5ml of ready-to-use probe reagent solutions.