Order Entry
Belgium
ContactUsLinkComponent
 
Wash buffer salts, lyophilised, Enzo Life Sciences
  ENZOENZ33802
 :  
Wash buffer salts, lyophilised, Enzo Life Sciences
  ENZOENZ33802
 :  

 

  • Pk:
    3 L
  • Description:
    Wash buffer salts, pack of 3
  • Seq:
    02
  • Formulation:
    Lyophilised
  • Sensitivity:
    Dissolve entire contents of packet in 1 L of distilled or deionised water. The pH of the solution should be 7,1 ±0,1.
  • Size:
    3 L

 

 

Wash Buffer Salts can be used in tissue preparations, immunohistochemical staining procedures and in assays that require HRP-linked detection reagents.

  • Each packet of salts yields 1 liter of buffered saline
  • pH should be 7.1 ± 0.1