ContactUsLinkComponent
AMPIGENE® Taq DNA polymerase
AMPIGENE® Taq DNA polymerase
Catalog # ENZOENZPRT1000500
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
AMPIGENE® Taq DNA polymerase
Catalog # ENZOENZPRT1000500
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    AMPIGENE® Taq DNA polymerase
  • Seq:
    02
  • Units:
    500
Specifications
About this item
Optimised standard PCR for enhanced speed, yield, and specificity.
AMPIGENE® Taq DNA Polymerase uses the latest developments in polymerase technology and buffer chemistry to optimize PCR. The AMPIGENE® Taq DNA Polymerase and 5x AMPIGENE® reaction buffer are separate for flexibility and end-user customization. It can be used for routine PCR applications including genotyping, screening, and library construction.
Ordering information: Contains AMPIGENE® Taq DNA polymerase (5u/µl)and 5x AMPIGENE® reaction buffer (15mM MgCl2, 5mM dNTPs, enhancers and stabilizers).