Order Entry
Belgium
ContactUsLinkComponent
Cyanine 5-dUTP
Cyanine 5-dUTP
Catalog # ENZOENZ42502
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Cyanine 5-dUTP
Catalog # ENZOENZ42502
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    25 Nmo
  • Description:
    Cyanine 5-dUTP 25 nmol
  • Seq:
    02
  • Purity detail:
    Purified by ion-exchange chromatography
  • Purity:
    ≥93% (HPLC)
  • Concentration:
    1 mM
  • Formulation:
    Liquid. Solution in water.

Specifications

About this item

Cyanine 5-dUTP can replace TTP in reactions in which it serves as a substrate for E. coli DNA polymerase (holoenzyme and Klenow fragment), T4 and Taq DNA polymerases, reverse transcriptase (from AMV and M-MuLV) and terminal transferase. Fluorescently labeled probes can be prepared with this fluorescent nucleotide by a variety of methods including nick translation, cDNA labeling and 3’-end labeling. Probes generated by these methods are suitable for use for the identification of specific sequences by in situ hybridization procedures on fixed cells and tissues by direct fluorescence detection. Cyanine 5-dUTP can also be used for multicolor fluorescence labeling.