Order Entry
Belgium
ContactUsLinkComponent
Blur 8 BioProbe®
Blur 8 BioProbe®
Catalog # ENZOENZ40849
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Blur 8 BioProbe®
Catalog # ENZOENZ40849
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    2 µG
  • Application:
  • Description:
    Blur 8 BioProbe®
  • Seq:
    02
  • Environmentally Preferable:
  • Concentration:
    20 µg/ml
  • Formulation:
    Liquid. In 10 mM TRIS HCl, pH 7,5, containing 1 mM EDTA.
  • Size:
    2 µg

Specifications

About this item

The Blur 8 BioProbe® labeled probe is prepared by nick translation of a clone of the human alu repeat sequence. Fragment size range: 100-1000 base pairs (as estimated by agarose gel electrophoresis).