Order Entry
Belgium
ContactUsLinkComponent
PROTEOSTAT® protein refolding and aggregation sensing kit for microplates
PROTEOSTAT® protein refolding and aggregation sensing kit for microplates
Catalog # ENZOENZ51040KP002
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
PROTEOSTAT® protein refolding and aggregation sensing kit for microplates
Catalog # ENZOENZ51040KP002
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    PROTEOSTAT® Protein refolding and aggregation sensing kit
  • Seq:
    02
  • Storage temperature:
    –20 °C
  • Tests per kit:
    192 reactions (2 plates)

Specifications

About this item

ProteoStat® Protein refolding and aggregation sensing kit provides a fractional factorial matrix design to facilitate screening of refolding parameters for a specific protein in different buffers, and a simple, homogenous assay format for monitoring protein aggregation using a red-emitting aggregation-sensitive dye. 

Caution: RUO - Research Use Only.