Caspase-1 colorimetric assay kit
Catalog #
Product Details & Documents
Based on the spectrophotometric detection of the chromophore pNA (p-nitroaniline) after cleavage from the labelled substrate YVAD-pNA.
Delivery information: Kit includes cell lysis buffer (25 ml), 2X reaction buffer(2 ml), 1 mM YVAD-pNA (125 µl), 1 M DTT (100 µl), dilution bBuffer (25 ml).