Order Entry
Belgium
ContactUsLinkComponent
Matrix Metalloproteinase-9 (MMP-9) colorimetric drug discovery kit
Matrix Metalloproteinase-9 (MMP-9) colorimetric drug discovery kit
Catalog # ENZOBMLAK4100001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Matrix Metalloproteinase-9 (MMP-9) colorimetric drug discovery kit
Catalog # ENZOBMLAK4100001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Matrix Metalloproteinase-9 (MMP-9) colorimetric drug discovery kit
  • Seq:
    02
  • Storage conditions:
    −80 °C
  • Application:
    Colorimetric detection, HTS activity assay
  • Size:
    1 kit

Specifications

About this item

A QuantiZyme™ Assay System. The MMP-9 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-9 (MMP-9, gelatinase B) using a thiopeptide as a chromogenic substrate.

Delivery: Kit contains MMP-9 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.