Order Entry
Belgium
ContactUsLinkComponent
Matrix Metalloproteinase-12 (MMP-12) colorimetric drug discovery kit
Matrix Metalloproteinase-12 (MMP-12) colorimetric drug discovery kit
Catalog # ENZOBMLAK4020001
Supplier:  Enzo Life Sciences
CAS Number:  
Matrix Metalloproteinase-12 (MMP-12) colorimetric drug discovery kit
Catalog # ENZOBMLAK4020001
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Matrix Metalloproteinase-12 (MMP-12) colorimetric drug discovery kit
  • Seq:
    02
  • Storage conditions:
    −80 °C
  • Application:
    Colorimetric detection, HTS activity assay
  • Size:
    1 kit

Specifications

About this item

A QuantiZyme™ Assay System. The MMP-12 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-12 (MMP-12, macrophage elastase, metalloelastase) using a thiopeptide as a chromogenic substrate. The assays are performed in a convenient 96-well plate format. Included are active enzyme, assay buffer, a prototypic control inhibitor (NNGH), and a detailed instruction booklet.

Delivery: Kit contains MMP-12 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH), Assay Buffer, 96-well Microplate and a detailed instruction booklet.