Order Entry
Belgium
ContactUsLinkComponent
 
Melatonin ELISA kit
Catalog # ENZOENZKIT1500001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Melatonin ELISA kit
Catalog # ENZOENZKIT1500001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Target protein:
    Melatonin
  • Description:
    Melatonin ELISA kit, grow cells
  • Seq:
    01
  • Sample type:
    Saliva, serum, plasma, and fruit homogenates
  • Time to Results:
    2 h
  • Sensitivity:
    0,162 ng/ml (0,08 - 50 ng/ml)

Specifications

About this item

Ultra-sensitive Melatonin ELISA kit enabling detection of Melatonin in saliva, serum, plasma, and fruit homogenates in just 2 hours.

  • Sensitive measurement of Melatonin, detecting as little as 0,162 ng/ml
  • Broad dynamic range suitable for a large variety of samples
  • High throughput format with results in 2 hours for up to 40 samples in duplicate
  • Validated for a variety of sample matrices including fruit homogenates

For the quantitative determination of melatonin in any species. Validated specifically for saliva, serum, plasma, and fruit samples.
The Melatonin ELISA kit is a colorimetric competitive immunoassay kit with results in 2 hours.

Ordering information: Contains Goat anti-mouse IgG Microtiter Plate, Assay Buffer 10, Standard, Wash Buffer Concentrate, Antibody, Tracer, Conjugate, TMB Substrate, and Stop Solution.