ACTH ELISA kit, grow cells
Catalog # ENZOENZKIT1380001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Target protein:Adrenocorticotropic Hormone (ACTH)
- Description:ACTH ELISA kit, grow cells
- Seq:01
- Sensitivity:0,46 pg/ml (0,5 - 165 pg/ml)
Specifications
About this item
Highly sensitive ACTH ELISA kit enabling detection of ACTH in human plasma in 4.5 hours.
- Sensitive measurement of ACTH, detecting as little as 0,46 pg/ml.
- Negligible reactivity to similar hormones.
- High throughput with results in 4.5 hours.
- Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of ACTH in plasma in human samples.
The ACTH ELISA kit is a colorimetric immunoassay kit with results in 4,5 hours.