Order Entry
Belgium
ContactUsLinkComponent
Peptide YY ELISA kit
Peptide YY ELISA kit
Catalog # ENZOENZKIT1330001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Peptide YY ELISA kit
Catalog # ENZOENZKIT1330001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Peptide YY ELISA kit, grow cells
  • Seq:
    01
  • Sensitivity:
    18,75 pg/ml

Specifications

About this item

Highly sensitive Peptide YY ELISA kit enabling detection of PYY in serum in 3,5 hrs.

  • Sensitive measurement of PYY, detecting as little as 18,75 pg/ml
  • Negligible reactivity from similar peptides
  • High throughput format with results in 3,5 hrs
  • Fully quantitative results that surpass semi-quantitative Western blot analysis

For the quantitative determination of Peptide YY in serum, plasma, cell culture supernatant and tissue homogenate in human samples.

The Peptide YY ELISA kit is a colorimetric immunometric immunoassay kit with results in 3,5 hours.

Ordering information: Contains Microtiter Plate, Standard, Sample Diluent, Detection Antibody, Wash Buffer Concentrate, Conjugate, Conjugate Diluent, Substrate, and Stop Solution.