NGAL (Monkey) ELISA kit, grow cells
Catalog # ENZOBPDKIT045
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Target protein:NGAL
- Description:NGAL (Monkey) ELISA kit, grow cells
- Seq:02
- Sensitivity:1,5 pg/ml (range 10 - 200 pg/ml)
Specifications
About this item
For the quantitative determination of NGAL in monkey urine, plasma, serum, tissue extracts, or culture supernatants.
- Ready-to-use: Pre-coated plate and individual calibrators save time and reduce errors
- Easy-to-use: Convenient storage at 4°C with room temperature incubations
- Rapid: Results in just four hours from up to 40 samples in duplicate
For the quantitative determination of NGAL in monkey urine, plasma, serum, tissue extracts, or culture supernatants.
A colorimetric immunometric ELISA kit for the rapid measurement of NGAL in pig samples. Absorbance is read at 450 nm.
Ordering information: Contains Microtiter Plate, Sample Diluent Conc., Monkey NGAL Calibrator Stock, Wash Solution Conc., Biotinylated Monkey-NGAL Antibody, HRP-Streptavidin, TMB Substrate, Stop Solution. Manufactured by BioPorto Diagnostics.