Order Entry
Belgium
ContactUsLinkComponent
Oxytocin ELISA kit
Oxytocin ELISA kit
Catalog # ENZOADI900153A0001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Oxytocin ELISA kit
Catalog # ENZOADI900153A0001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Target protein:
    Oxytocin
  • Description:
    Oxytocin ELISA kit
  • Seq:
    02
  • Sensitivity:
    15 pg/ml (15,6 - 1,000 pg/ml)
  • Tests per kit:
    96 Wells

Specifications

About this item

The most sensitive, specific and flexible alternative to more costly and time-consuming methods for oxytocin detection.

For the quantitative determination of oxytocin in culture supernatants, milk, plasma, and serum from any species. Cited sample types include cerebral spinal fluid, saliva, tissue and urine.

Ordering information: Contains GxR IgG microtiter plate, conjugate, antibody, assay buffer, wash buffer concentrate, standard, pNpp substrate, stop solution. Alternative size available: ADI-901-153A (5 x 96 wells).