Order Entry
Belgium
ContactUsLinkComponent
Anti-HSP70 IgG/A/M (human), ELISA Kit
Anti-HSP70 IgG/A/M (human), ELISA Kit
Catalog # ENZOADIEKS750
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Anti-HSP70 IgG/A/M (human), ELISA Kit
Catalog # ENZOADIEKS750
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Human
  • Description:
    Anti-HSP70 IgG/A/M (human), ELISA Kit
  • Seq:
    02
  • Sensitivity:
    6,79 ng/ml (range 31,25 - 1,000 ng/ml)

Specifications

About this item

Most comprehensive portfolio of heat shock protein (chaperone) products commercially available.

For the quantitative determination of anti-HSP70 IgG/A/M autoantibodies in human serum.

This assay allows for reproducible, accurate and precise determination of IgG , IgA and IgM antibodies (total) to human Hsp70 in serum. The ELISA kit uses recombinant human Hsp70 bound to the wells of the plate to bind anti-human Hsp60 antibodies present in human serum. The captured anti-human Hsp70 antibodies are detected with a HRP conjugated goat polyclonal antibody specific for human IgG, IgA and IgM molecules. The assay is developed with tetramethylbenzidine substrate pro ducing a blue color in proportion to the amount of captured anti -human Hsp70 antibodies. The color development is stopped with acid stop solution which converts the endpoint color to yellow. The intensity of the color is measured in a microplate reader at 450 nm.

Ordering information: Contains Microtiter plate, Standard, Sample diluent 2, Wash buffer concentrate, HRP Conjugate, TMB Substrate, Stop solution 2.