Order Entry
Belgium
ContactUsLinkComponent
PGE2 high sensitivity ELISA kit, plasma
PGE2 high sensitivity ELISA kit, plasma
Catalog # ENZOADI930001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
PGE2 high sensitivity ELISA kit, plasma
Catalog # ENZOADI930001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Target protein:
    PGE2
  • Description:
    PGE2 high sensitivity ELISA kit, plasma
  • Seq:
    02
  • Sensitivity:
    8,26 pg/ml (range 7,8 - 1,000 pg/ml)

Specifications

About this item

Ultrasensitive PGE2 ELISA kit for inflammation and eicosanoid research.

For the quantitative determination of PGE2 in culture supernatants, serum, saliva, urine, and whole blood from any species. Cited sample type includes plasma.

The PGE2 high sensitivity EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 1 hour.

Ordering information: Contains GxM IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-766 (5x96 wells).