Order Entry
Belgium
ContactUsLinkComponent
ImmunoSet® αB-Crystallin ELISA development set
ImmunoSet® αB-Crystallin ELISA development set
Catalog # ENZOADI960074
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
ImmunoSet® αB-Crystallin ELISA development set
Catalog # ENZOADI960074
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 SET
  • Assay duration:
    Multiple steps
  • Format:
    Uncoated
  • Primary antibody reactivity:
    Human
    Bovine
    Rat
    Mouse
  • Target protein:
    αB-Crystallin
  • Description:
    ImmunoSet® αB-Crystallin ELISA development set
  • Seq:
    02
  • Detection method:
    Colorimetric 450 nm
  • Sensitivity:
    0,59 ng/ml (range 1,25 - 40 ng/ml)
  • Tests per kit:
    5×96 wells

Specifications

About this item

For the quantitative determination of αB-Crystallin in cell lysates of human, mouse, rat, and cow origin.

The ImmunoSet® αB-Crystallin EIA development set contains sufficient reagents for the development of 5×96 well EIA plates which provides analysis of ~200 samples in duplicate. Save time, money, and sample by developing your own EIA kits. Simply coat your plates and go! This kit provides superior accuracy with fully quantitative results compared to Western blot analysis and has low reactivity with related molecules.

Ordering information: Contains Capture antibody, Standard, Detection antibody, SA-HRP.