Order Entry
Belgium
ContactUsLinkComponent
IgM (mouse), ELISA kit
IgM (mouse), ELISA kit
Catalog # ENZOADI900120
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
IgM (mouse), ELISA kit
Catalog # ENZOADI900120
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    IgM
  • Description:
    IgM (mouse), ELISA kit
  • Seq:
    02
  • Sensitivity:
    0,60 ng/ml (range 3,91 - 250 ng/ml)

Specifications

About this item

For the quantitative determination of IgM in ascites fluid, culture supernatants, and serum of mouse origin.

The IgM (mouse), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 90 minutes. Absorbance is read at 450 nm. Ideal for determining the isotype and concentration of monoclonal antibodies.

Ordering information: Contains GxM IgG Microtiter plate, mouse IgM conjugate, Assay buffer 13 concentrate, Wash buffer concentrate, mouse IgGM standard, TMB Substrate, Stop solution 2.