Complement C3a des Arg (human), ELISA kit
Catalog # ENZOADI900058
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Assay Type:Competitive
- Primary antibody reactivity:Human
- Description:Complement C3a des Arg (human), ELISA kit
- Seq:02
- Sensitivity:0,120 ng/ml (range 0,313 - 20 ng/ml)
- Tests per kit:96 wells
Specifications
About this item
For the quantitative determination of Complement C3a des arg in human EDTA plasma.
The Complement C3a des Arg (human), EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. This kit comes ready-to-use with sample preparation reagents included. The Complement C3a des Arg (human), EIA kit is specific, offering low cross-reactivity to structurally related molecules, and sensitive with quantification of as little as 120pg/ml Complement C3a des Arg.
Ordering information: Contains GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer 10 concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Complement reagent A, Complement reagent B.