Order Entry
Belgium
ContactUsLinkComponent
Leptin (human), ELISA kit
Leptin (human), ELISA kit
Catalog # ENZOADI900028A
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Leptin (human), ELISA kit
Catalog # ENZOADI900028A
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Human
  • Target protein:
    Leptin
  • Description:
    Leptin (human), ELISA kit
  • Seq:
    02
  • Sensitivity:
    23,4 pg/ml (range 31,3 - 2000 pg/ml)

Specifications

About this item

For the quantitative determination of human leptin in culture supernatants, plasma, and serum.

The Leptin (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours.

Ordering information: Contains Microtiter plate, Standard, Antibody, Conjugate, Assay buffer 17, TMB Substrate, Stop solution 2, Wash buffer concentrate.