Order Entry
Belgium
ContactUsLinkComponent
TXB2 ELISA Kits
TXB2 ELISA Kits
Catalog # ENZOADI900002
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
TXB2 ELISA Kits
Catalog # ENZOADI900002
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Primary antibody reactivity:
    Multiple
  • Target protein:
    TXB2
  • Description:
    TXB2 ELISA kit
  • Seq:
    02
  • Sensitivity:
    10,54 pg/ml (range 13,7 - 10,000 pg/ml)

Specifications

About this item

For the quantitative determination of TXB2 in culture supernatants, plasma, serum, saliva, and urine from any species. Cited sample types include cell lysate, coronary effluent, liver perfusate, platelets, rectal dialysate, and whole blood.

The TXB2 ELISA kit is a colorimetric competitive enzyme immunoassay kit with results in 2 in a convenient 96 well plate based assay. This kit is highly cited in peer-reviewed publications in a diverse assortment of sample types including mucosa, tissue, serum, plasma, urine, and much more. The TXB2 ELISA kit provides easy-to-use stable liquid color-coded reagents that reduce errors.

Ordering information: Contains GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-270 (5x96 wells)