Order Entry
Belgium
ContactUsLinkComponent
 
Testosterone ELISA kit
Catalog # ENZOADI901065
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Testosterone ELISA kit
Catalog # ENZOADI901065
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Target protein:
    Testosterone
  • Description:
    Testosterone ELISA kit
  • Seq:
    02
  • Time to Results:
    3 h
  • Sensitivity:
    5,67 pg/ml (Range 7,81 - 2000 pg/ml)

Specifications

About this item

For the quantitative determination of testosterone in culture supernatants, plasma, serum, and saliva from any species. Not suitable for human serum and plasma. For those samples use product 89141-348. Cited sample type includes tissue and feces.

The Testosterone EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. Ready-to-use liquid color-coded reagents are provided to reduce error. The reproducible results lot-after-lot provide excellent consistency, ideal for longitudinal studies.

Ordering information: Contains GxM Microtiter plate, Conjugate, Antibody, Assay buffer 3, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Steroid displacement reagent. Alternative size available: 89141-112 (1x96 wells).