Order Entry
Belgium
ContactUsLinkComponent
8-iso-PGF2α ELISA kits
8-iso-PGF2α ELISA kits
Supplier:  ENZO LIFE SCIENCES
Certificates
8-iso-PGF2α ELISA kits
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Assay Type:
    Competitive
  • Conjugate ELISA:
    Alkaline Phosphatase (AP)
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Species independent
  • Target protein:
    8-iso-PGF2α
  • Detection method:
    Colorimetric detection 405 nm
  • Time to Results:
    <3 h
  • Sensitivity:
    16,3 pg/ml (range 6,1 - 100000 pg/ml)

Specifications

Frequently Bought Together