ADMA (human) ELISA kit
Catalog # ENZOALX850323KI01
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Human
- Target protein:ADMA
- Description:ADMA (human) ELISA kit
- Seq:02
- Sensitivity:0,05 μmol/L (expected value: 0,45 µmol/L ± 0,19 µmol/L)
Specifications
About this item
For the quantitative determination of asymmetrical dimethylarginine (ADMA) in human serum, citrate- and EDTA-plasma. Does not cross-react with SDMA, NMMA or L-arginine.