Order Entry
Belgium
ContactUsLinkComponent
6-keto-PGF1α ELISA Kits
6-keto-PGF1α ELISA Kits
  ENZOADI901004
 :  
6-keto-PGF1α ELISA Kits
  ENZOADI901004
 :  

 

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Target protein:
    6-keto-PGF1α
  • Description:
    6-keto-PGF1α ELISA kit
  • Seq:
    02
  • Sensitivity:
    1,40 pg/ml (range 3,2 - 50,000 pg/ml)

 

 

For the quantitative determination of 6-keto-PGF1α in culture supernatants, serum, saliva, and urine from any species.

The 6-keto-PGF1α EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 1α EIA kit has been fully validated in-house in a wide variety of sample matrices.

 : Contains DxS Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-056 (1x96 wells).