Order Entry
Belgium
ContactUsLinkComponent
Protein carbonyl ELISA kit
Protein carbonyl ELISA kit
Catalog # ENZOALX850312KI01
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Protein carbonyl ELISA kit
Catalog # ENZOALX850312KI01
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Protein carbonyl ELISA kit
  • Seq:
    02

Specifications

About this item

The Protein carbonyl ELISA kit is an antibody-based detection set. It has higher sensitivity, lower background, is less labour-intensive and handles more samples per day as compared to colorimetric assays.

  • Detecting low levels of carbonylated proteins
  • Validated in various sample types
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
  • Assay Time: 4,5 hours

For quantitative determination of carbonylated protein levels in plasma, other body fluids, cell and tissue extracts.