PTX3 (human) detection set [for ELISA application]
Catalog # ENZOALX850299KI01
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 SET
- Primary antibody reactivity:Human
- Target protein:PTX3
- Description:PTX3 (human) detection set [for ELISA application]
- Seq:02
- Storage conditions:−20 °C (long term)
Specifications
About this item
For the quantitative determination of PTX3 in human and bovine (weak) cell culture media, EDTA-plasma or serum (weak) samples.
Delivery: Kit includes 1 vial (10 µg) PTX3 (human), 1 vial (50 µg) PTX3, pAb (Biotin), 1 vial (100 µg) PTX3, mAb (MNB4) for preparing 10 plates.