Order Entry
Belgium
ContactUsLinkComponent
PMN-Elastase (human) ELISA kit
PMN-Elastase (human) ELISA kit
Catalog # ENZOALX850265KI01
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
PMN-Elastase (human) ELISA kit
Catalog # ENZOALX850265KI01
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Human
  • Description:
    PMN-Elastase (Human) ELISA kit
  • Seq:
    02
  • Storage conditions:
    4 °C
  • Sensitivity:
    0,06 ng/mL (Range 0,16 - 10 ng/ml)

Specifications

About this item

For the quantitative determination of human PMN-elastase in cell culture supernatants, plasma and other body fluids.

  • Assay Time: 3,5 hours