Cellular senescence activity assay
Catalog # ENZOENZKIT1290120
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:120 Assays
- Description:Cellular senescence activity assay
- Seq:01
Specifications
About this item
Highly reproducible SA-β-gal activity kit
- Quantify SA-β-gal activity in a fluorescence plate reader
- Easy-to-use protocol with results in less than 90 minutes
- Fully compatible with all cell types
For the determination of cellular senescence by measuring SA-β-gal activity using a fluorometric substrate. The 96-well Cellular senescence assay kit provides an easy-to-use and efficient method to determine the cellular senescence by measuring SA-β-gal activity using a fluorometric substrate. This quantitative assay uses cell lysate for both SA-β-galactosidase activity determination and normalization of samples containing different cell numbers. Each kit provides sufficient quantities to perform up to 120 assays in a 96-well plate.
Ordering information: Contains 2X Cell lysis buffer, 2X Reaction buffer, SA-?-gal substrate (20X), Stop solution.