Cellular senescence live cell analysis assay
Catalog # ENZOENZKIT1300010
Fournisseur: Enzo Life Sciences
Numéro CAS:
Spécifications
- Pk:10 Assays
- Description:Cellular senescence live cell analysis assay
- Seq:01
Spécifications
À propos de cet article
Simple assay to quantify senescence-associated β-galactosidase activity, in live cells, using flow cytometry or fluorescent microscopy.
For the study of cellular senescence in live cells.
Assay kit that uses a fluorogenic substrate to measure SA-β-gal activity in live cells.
Informations de commande: Contains Cell pretreatment solution (1000x) and SA-?-gal Substrate (200x).