Order Entry
Belgium
ContactUsLinkComponent
Mito-ID® Membrane potential cytotoxicity kit for microplates
Mito-ID® Membrane potential cytotoxicity kit for microplates
Catalog # ENZOENZ51019KP002
Supplier:  Enzo Life Sciences
CAS Number:  
Mito-ID® Membrane potential cytotoxicity kit for microplates
Catalog # ENZOENZ51019KP002
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    MITO-ID® Membrane potential cytotoxicity kit
  • Seq:
    02
  • Size:
    1 kit
  • Tests per kit:
    2× 96-well microplates

Specifications

About this item

A Real-time Mitochondrial membrane Potential Assay with Superior Sensitivity. Suitable for high-throughput applications.

  • 10X more sensitive than JC-1 with superior aqueous solubility
  • Photostable dual-emission dye
  • Suitable for time-course studies evaluating intact and compromised mitochondria
  • No-wash/No-medium removal
  • Separate MITO-ID® assay is available for detection of mitochondrial mass
  • Detects toxicity at lower drug/dose concentrations
  • No solvent artifacts as those seen with JC-1 formulation

MITO-ID® Membrane Potential Cytotoxicity Kit enables monitoring of mitochondrial potential changes using a simple fluorescence microplate reader.

The Mito-ID® Membrane Potential Cytotoxicity Kit measures fluctuations in mitochondrial membrane potential (MMP) utilizing a cationic dual-emission dye that exists as green fluorescent monomers in the cytosol, and accumulates as orange fluorescent J-aggregates in the mitochondria. Mitochondria having a low membrane potential will accumulate low concentrations of dye and will exhibit green fluorescence while more highly polarized mitochondria will exhibit orange-red fluorescence. Cells exhibit a shift from orange to green fluorescence as mitochondrial function becomes increasingly compromised. The kit is a unique HTS assay that monitors mitochondrial membrane potential in real-time without the need for washes or medium removal.

Mechanism of Action

The basic chemical structure of the dye consists of highly conjugated moieties that extensively delocalize a positive charge thus allowing electrophoretic uptake toward the negatively charged matrix phase of the polarized inner mitochondrial membrane. The dye is capable of entering selectively into mitochondria wherein it changes its color reversibly from green to orange as membrane potential increases (dual-emission potential probe). This photophysical property is due to the reversible formation of J-aggregates upon membrane polarization that causes shifts in emitted light from ~530nm (the emission of the monomeric dye) to 590 nm (the emission of the J-aggregate form) when excited at 490 nm. As a consequence, mitochondria having a low membrane potential will accumulate low concentrations of dye and will exhibit green fluorescence while more highly polarized mitochondria will exhibit orange fluorescence.

Ordering information: Quantity: For 2 x 96-well microplates Contents: MITO-ID® MP Detection Reagent, 200 µl
CCCP Control, 100 µl
10X Assay Buffer 1: 2,5 ml
50X Assay Buffer 2: 0,5 ml