Order Entry
Belgium
ContactUsLinkComponent
Apoptosis/Necrosis Detection Kits, for microscopy and flow cytometry, GFP-Certified™
Apoptosis/Necrosis Detection Kits, for microscopy and flow cytometry, GFP-Certified™
Supplier:  ENZO LIFE SCIENCES
Certificates
undefined
Apoptosis/Necrosis Detection Kits, for microscopy and flow cytometry, GFP-Certified™
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Storage conditions:
    4 °C
  • Application:
    Flow Cytometry, Fluorescence Microscopy, Fluorescent Detection
  • Size:
    1 kit

Specifications

Frequently Bought Together