Order Entry
Belgium
ContactUsLinkComponent
 
OXI-TEK TBARS assay kit
Catalog # ENZOALX850287KI01
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
OXI-TEK TBARS assay kit
Catalog # ENZOALX850287KI01
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    160 Tests
  • Description:
    OXI-TEK TBARS assay kit
  • Seq:
    02
  • Storage temperature:
    4 °C (long term)
  • Application:
    Fluorescence Microscopy
  • Size:
    160 tests

Specifications

About this item

The sensitivity of measuring thiobarbituric acid reactive substances (TBARS) has made this assay the method of choice for screening and monitoring lipid peroxidation, which is a major indication of oxidative stress.

Malondialdehyde (MDA) forms a 1:2 adduct with thiobarbituric acid; the adduct can be measured by fluorometry. Biological specimens contain a mixture of TBARS, including lipid hydroperoxides and aldehydes. In practice, TBARS are expressed in equivalent numbers to MDA.