Cytochrome C releasing apoptosis assay kit
N° de catalogue ENZOALX850277KI01
Fournisseur: Enzo Life Sciences
Numéro CAS:
Spécifications
- Pk:1 KIT
- Description:Cytochrome c releasing apoptosis assay kit
- Seq:02
- Storage conditions:–20 °C (long term)
- Size:1 kit, 100 tests
- Tests per kit:100
Spécifications
À propos de cet article
For the detection of Cytochrome c translocation from mitochondria into cytosol during apoptosis.
The kit provides an easy-to-use procedure and reagents for separating a mitochondria-enriched fraction from cytosol. Cytochrome c released from mitochondria into cytosol is determined by Western blot using the cytochrome c antibody provided in the kit. The procedure is simple, straightforward and requires no ultracentrifugation.
Livraison: Kit includes Mitochondria Extraction Buffer (10 mL), 5X Cytosol Extraction Buffer (20 mL), 1 M DTT (110 µL), 500X Protease Inhibitor Cocktail (1 vial), 0.2 mg/mL Cytochrome c Antibody (0.5 mL).