Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Assays
Enzyme Assays
Cyclic ucleotide phosphodiesterase assay kit components
Cyclic ucleotide phosphodiesterase assay kit components
Cyclic ucleotide phosphodiesterase assay kit components
Catalog # ENZOBMLKI1900002
Supplier:
Enzo Life Sciences
CAS Number:
Expand image
Cyclic ucleotide phosphodiesterase assay kit components
Catalog # ENZOBMLKI1900002
Supplier:
Enzo Life Sciences
CAS Number:
Specifications
Pk:
2 mL
Description:
3', 5'-cGMP substrate
Seq:
08
Specifications
About this item
Product Details
Product Details