Order Entry
Belgium
ContactUsLinkComponent
Tween-20 solution (10%)
Tween-20 solution (10%)
Catalog # ENZOADI801929
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Tween-20 solution (10%)
Catalog # ENZOADI801929
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    30 mL
  • Description:
    Tween-20 solution, 10%
  • Seq:
    02

Specifications

About this item

This Tween®-20 solution (10%) is specially designed to create the Blocking Buffer and Assay Buffer in ImmunoSet products. Each bottle is sufficient for use with 5×96 wells microtiter plates if following the instructions in our ImmunoSet protocol.

  • Long Term Storage: 4 °C

The Tween-20 solution may be used for any other assay development needs. Additional assay development reagents are available as companion products to our ImmunoSet products.