Specifications
- Antibody type:Primary
- Antigen name:Early Endosome Antigen 1
- Antigen symbol:EEA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Seq:02
- Form:Liquid
- Antigen synonyms:ZFYVE2|MSTP105|MST105
- Amino acid number:1391 to 1410
- Storage buffer:PBS containing 1 mg/ml BSA and 0,05% sodium azide.
- Sequence:TPSSKKPVRVCDACFNDLQG
- Storage temperature:–20 °C
- Shipping temperature:Shipped on blue ice.
- Immunogen:Synthetic peptide corresponding to aa 1391-1410 (T1391PSSKKPVRVCDACFNDLQG1410) of human EEA1 (early endosomal antigen 1).
- Purification:Purified by epitope affinity chromatography
- Pk:100 µG
Specifications
About this item
EEA1 is a 162kDa membrane bound protein component specific to the early endosome and is essential for fusion between early endocytic vesicles. Early endosomes are cytoplasmic compartments which fuse with endocytic vesicles for redistribution of extracellular compounds to alternate destinations. Zinc-finger-like domains reminiscent of those found in nucleic acid binding proteins are located in the amino and carboxyl-terminal domains of EEA1. The COOH-terminal zinc-finger-like-domain is conserved in several other non-nuclear proteins, some of which are also involved in intracellular protein trafficking. In addition, this domain is an authentic zinc-binding domain and is critical to the intracellular localization of EEA1. Membrane association of EEA1 has been shown to be dependent on phosphatidyl 3-kinase activity, inhibitors of which cause EEA1 to dissociate from early endosomes.
Type: Primary
Antigen: EEA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse