Order Entry
Belgium
ContactUsLinkComponent
Anti-HSP90A Mouse Monoclonal Antibody [clone: K41009]
Anti-HSP90A Mouse Monoclonal Antibody [clone: K41009]
Supplier:  Enzo Life Sciences
Certificates
Anti-HSP90A Mouse Monoclonal Antibody [clone: K41009]
Supplier:  Enzo Life Sciences

Specifications

  • Catalog No:
  • ENZOADISPA839050
  • ENZOADISPA839200
  • Antigen symbol:
  • HSP90A
  • HSP90A
  • Antigen name:
  • Heat Shock Protein 90 alpha
  • Heat Shock Protein 90 alpha
  • Antigen synonyms:
  • HSP84|HSP90-BETA|heat shock 90kDa protein 1|heat shock 90kD protein|D6S182|HSPCB|HSPC2|HSPCA|heat shock protein HSP 90-beta|alpha|heat shock 90kD protein 1|heat shock 84 kDa|Hsp89|HSP86|renal carcinoma antigen NY-REN-38|HSP 86|heat shock 86 kDa|HSPCAL1|HSP 90|HSP90B|HSPN|Hsp90|beta|alpha-like 4|HSPC1|heat shock protein HSP 90-alpha|90-kda heat shock protein beta HSP90 beta|HSP90N|HSPCAL4|Hsp90aa1|LAP2|HSP89A|HSP90A|HSP 84|HSP90AB1|heat shock protein beta
  • HSP84|HSP90-BETA|heat shock 90kDa protein 1|heat shock 90kD protein|D6S182|HSPCB|HSPC2|HSPCA|heat shock protein HSP 90-beta|alpha|heat shock 90kD protein 1|heat shock 84 kDa|Hsp89|HSP86|renal carcinoma antigen NY-REN-38|HSP 86|heat shock 86 kDa|HSPCAL1|HSP 90|HSP90B|HSPN|Hsp90|beta|alpha-like 4|HSPC1|heat shock protein HSP 90-alpha|90-kda heat shock protein beta HSP90 beta|HSP90N|HSPCAL4|Hsp90aa1|LAP2|HSP89A|HSP90A|HSP 84|HSP90AB1|heat shock protein beta
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • K41009
  • K41009
  • Reactivity:
  • Rabbit, Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG2a
  • IgG2a
  • Western blot:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant human Hsp90alpha
  • Recombinant human Hsp90alpha
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Purified by Protein G affinity chromatography
  • Purified by Protein G affinity chromatography
  • Storage buffer:
  • PBS, pH 7,2, containing 0,09% sodium azide
  • PBS, pH 7,2, containing 0,09% sodium azide
  • Storage temperature:
  • –20 °C
  • –20 °C
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together