Anti-Vasoactive Intestinal (Poly)Peptide Rabbit Polyclonal Antibody
Catalog #
Product Details & Documents
Host: Rabbit
- For Immunohistochemistry
Recognizes human, rat and pig VIP.
Type: Primary
Antigen: VASOACTIVE INTESTINAL (POLY)PEPTIDE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human



