Anti-CASP11 Rabbit Polyclonal Antibody
Catalog # ENZOBMLSA3240100
Supplier: Enzo Life Sciences
Specifications
- Antibody type:Primary
- Antigen name:Caspase 11
- Antigen symbol:CASP11
- Clonality:Polyclonal
- Host:Rabbit
- Western blot:Yes
- Seq:02
- Form:Liquid, antiserum containing sodium azide
- Antigen synonyms:CASP-11|ICEB,ICH-3
- Pk:100 µl
Specifications
About this item
Host: Rabbit
- For Western Blot
Recognizes mouse caspase-11.
Type: Primary
Antigen: CASP11
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse