Anti-CD42C Mouse Monoclonal Antibody
Catalog # ENZOALX805026C100
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen symbol:CD42C
- Clonality:Monoclonal
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Western blot:Yes
- Seq:02
- Form:Liquid. 100 µg of purified antibody in 200 µl PBS containing 0,05% sodium azide
- Purification:0.95
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Host: Mouse, Isotype: IgG2b
- For Immunoprecipitation, Western Blot
Recognizes the human GPIbbeta subunit (CD42c) of the human GPIb/IX/V (CD42a/CD42b/CD42c/CD42d) complex.
Type: Primary
Antigen: CD42C
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: