Anti-TICAM1 Rabbit Polyclonal Antibody
Catalog # ENZOALX215016R200
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen name:TIR Domain-containing Adapter Molecule 1
- Antigen symbol:TICAM1
- Clonality:Polyclonal
- Host:Rabbit
- Western blot:Yes
- Seq:02
- Form:Liquid. In PBS containing 0,05% BSA and 0,05% sodium azide.
- Antigen synonyms:MyD88-3|vinculin and TIR domain-containing protein B|PRVTIRB|Proline-rich|TRIF|IIAE6|TIR domain-containing adapter molecule 1|TICAM-1
- Pk:200 µl
Specifications
About this item
Host: Rabbit
- For Western Blot
Recognizes human and mouse TRIF.
Type: Primary
Antigen: TICAM1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human