Anti-CASP9 Rabbit Polyclonal Antibody
Catalog # ENZOALX210815C100
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen name:Caspase 9
- Antigen symbol:CASP9
- Clonality:Polyclonal
- Host:Rabbit
- Western blot:Yes
- Seq:02
- Form:Liquid. In PBS, pH 7,2, containing 0,5% BSA, 0,01% thimerosal and 30% glycerol.
- Antigen synonyms:EC 3.4.22.62|ICE-like apoptotic protease 6,ICE-LAP6,Apoptotic protease Mch-6,Apoptotic protease-activating factor 3|Apaf-3|CASP-9
- Concentration:0,2 mg/ml
- Purification:Epitope-affinity purified
- Pk:100 µG
Specifications
About this item
Host: Rabbit
- For Western Blot
Recognizes human, mouse, rat, hamster, rabbit, pig, dog, monkey and bovine caspase-9. Detects procaspase-9 and a caspase-9 cleavage product by Western blot.
Type: Primary
Antigen: CASP9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human