Anti-CASP1 Rabbit Polyclonal Antibody
Catalog # ENZOALX210804C100
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen name:Caspase 1
- Antigen symbol:CASP1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Seq:02
- Form:Liquid. In PBS, pH 7,2, containing 30% glycerol, 0,5% BSA and 0,01% thimerosal.
- Antigen synonyms:interleukin 1-B converting enzyme|ICE|IL1B-convertase|P45|apoptosis-related cysteine peptidase interleukin 1|IL1BC|beta|IL-1 beta-converting enzyme|caspase 1|interleukin 1|convertase|caspase-1|CASP1 nirs variant 1
- Concentration:0,2 mg/ml
- Purification:Epitope-affinity purified
- Pk:100 µG
Specifications
About this item
Host: Rabbit
- For Immunocytochemistry, Immunoprecipitation, Western Blot
Recognizes human, mouse and rat caspase-1. Detects bands of ~45kDa (procaspase-1) and ~20kDa (cleavage product) by Western blot.
Type: Primary
Antigen: CASP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human