Order Entry
Belgium
ContactUsLinkComponent
 
Anti-NNOS Rabbit Polyclonal Antibody
Catalog # ENZOALX210529C100
Supplier:  ENZO LIFE SCIENCES
Anti-NNOS Rabbit Polyclonal Antibody
Catalog # ENZOALX210529C100
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Nitric Oxide Synthase 1 (Neuronal)
  • Antigen symbol:
    NNOS
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid. In PBS containing 1 mg/ml BSA and 0,05% sodium azide.
  • Antigen synonyms:
    bNOS1|Neuronal Nitric Oxide Synthase|NOS Type 1|Brain NOS
  • Concentration:
    1 mg/ml
  • Purification:
    Epitope-affinity purified
  • Pk:
    100 µG

Specifications

About this item

Host: Rabbit

  • For Western Blot

Recognizes human nNOS. Detects a band of ~140kDa by Western blot.

Type: Primary
Antigen: NNOS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human