Order Entry
Belgium
ContactUsLinkComponent
Anti-SNAP25 Rabbit Polyclonal Antibody
Anti-SNAP25 Rabbit Polyclonal Antibody
Catalog # ENZOALX210279R100
Supplier:  ENZO LIFE SCIENCES
Anti-SNAP25 Rabbit Polyclonal Antibody
Catalog # ENZOALX210279R100
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Synaptosomal-Associated Protein, 25kDa
  • Antigen symbol:
    SNAP25
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid. In PBS containing 1 mg/ml BSA and 0,05% sodium azide.
  • Antigen synonyms:
    FLJ23079|synaptosomal-associated protein 25|SUP|SEC9|synaptosomal-associated 25 kDa protein|bA416N4.2|dJ1068F16.2|SNAP|SNAP-25|RIC-4|RIC4|resistance to inhibitors of cholinesterase 4 homolog|super protein
  • Purification:
    Epitope-affinity purified IgG
  • Pk:
    100 µl

Specifications

About this item

SNAP-25 (synaptosomal associated protein of 25kDa) is a soluble protein of 25kDa which plays a key role in vesicle membrane fusion events with the plasma membrane. This protein was originally discovered to be enriched in subsets of neurons in the brain and displays a presynaptic pattern of expression. The membrane proteins SNAP-25, synaptobrevin, and syntaxin form the core of a ubiquitous membrane fusion machine that interacts with the soluble proteins NSF and α-SNAP. Functional interactions have been demonstrated to exist between SNAP-25, syntaxin, and the synaptic protein interaction site on voltage-sensitive L- and N-type Ca2+ channels. SNAP-25 has been shown to be required for synaptic vesicle fusion with the plasma membrane during release of NGF. Regulated exocytosis of cortical granule secretion in fertilized eggs and membrane fusion events in neurons and endocrine cells is mediated by SNAP-25 in a Ca2+-dependent mechanism. SNAP-25 protein levels have been shown to be elevated in prolactinoma and GH/PRL tumor cells while reduced SNAP-25 protein expression has been observed between schizophrenic and normal hippocampal cells. Altered patterns of SNAP-25 expression have also been observed in the inferior temporal cortex and prefrontal association cortex between normal brains and brains from individuals affected with schizophrenia. SNAP-25 accumulation due to cytoskeletal dysfunction is also observed in the swollen axons of the white matter of individuals with severe Alzheimer's dementia.

  • For Immunocytochemistry, Western Blot

Recognizes mouse, rat, bovine and zebrafish SNAP-25. Detects a band of ~25kDa by Western blot.

Type: Primary
Antigen: SNAP25
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse