Order Entry
Belgium
ContactUsLinkComponent
Anti-RSEC8 Mouse Monoclonal Antibody
Anti-RSEC8 Mouse Monoclonal Antibody
Supplier:  ENZO LIFE SCIENCES
Certificates
Anti-RSEC8 Mouse Monoclonal Antibody
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Catalog No:
  • ENZOADIVAMSV016D
  • ENZOADIVAMSV016F
  • Antigen symbol:
  • RSEC8
  • RSEC8
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Reactivity:
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Form:
  • Liquid. In PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide.
  • Purification:
  • Protein G affinity purified

Specifications

Frequently Bought Together