Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Iron hematoxylin
Iron hematoxylin solution 1 Weigert's, Clin-Tech
Iron hematoxylin solution 1 Weigert's, Clin-Tech
Flammable
Health
Irritant
Iron hematoxylin solution 1 Weigert's, Clin-Tech
Catalog # VWRK640490
Supplier:
KLINIPATH
CAS Number:
SDS
Expand image
Flammable
Health
Irritant
Iron hematoxylin solution 1 Weigert's, Clin-Tech
Catalog # VWRK640490
Supplier:
KLINIPATH
CAS Number:
SDS
Specifications
Pk:
1 L
Seq:
5037
Specifications
About this item
Product Details
Documentation
:
UN:
1170
ADR:
3,II
:
GHS02
:
GHS08
:
GHS07
:
1
MDL Number:
MFCD00284879
Product Details
Documentation